Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010661.2 | NP_034791.2 | 5 | 1077 | ||
GenBank mRNA | AK053362.1 | - | 5 | 1078 | ||
AK143182.1 | - | 5 | 1076 | |||
BC131911.1 | - | 5 | 1073 | 40130188 | ||
U02880.1 | - | 5 | 1074 |