Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004855.4 | NP_004846.4 | 3 | 662 | ||
XM_011522235.2 | XP_011520537.1 | 3 | 662 | |||
XM_011522236.2 | XP_011520538.1 | 3 | 662 | |||
XM_017022730.1 | XP_016878219.1 | 1 | 26 | |||
GenBank mRNA | AK223330.1 | - | 3 | 367 | ||
AK314706.1 | - | 3 | 637 | |||
BC017711.1 | - | 3 | 599 | 4430974 | ||
D42138.1 | - | 3 | 390 |