Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Csad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144942.4 | NP_659191.1 | 4, 5 | 505 | ||
GenBank mRNA | AK005015.1 | - | 4, 5 | 506 | ||
AY033912.1 | - | 4, 5 | 422 |