Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem161a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293797.1 | NP_001280726.1 | 3 | 144 | ||
NM_145597.4 | NP_663572.1 | 4 | 248 | |||
GenBank mRNA | AK049979.1 | - | 4 | 247 | ||
AK082758.1 | - | 4 | 244 | |||
AK082916.1 | - | 3 | 145 | |||
AK088569.1 | - | 3 | 134 | |||
AK147962.1 | - | 4 | 241 | |||
AK180152.1 | - | 1 | 32 | |||
AK198557.1 | - | 2 | 47 | |||
AK215127.1 | - | 1 | 32 | |||
BC021367.1 | - | 4 | 248 | 5030434 |