Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D230025D16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145604.2 | NP_663579.2 | 5 | 561 | ||
GenBank mRNA | AK051965.1 | - | 5 | 547 | ||
AK156120.1 | - | 5 | 563 | |||
AK166937.1 | - | 4 | 1748 | |||
AK213296.1 | - | 3 | 120 | |||
AK218692.1 | - | 4 | 204 | |||
BC006874.1 | - | 5 | 444 | 2646515 | ||
BC057919.1 | - | 5 | 550 | 5134302 | ||
BC066141.1 | - | 5 | 482 | |||
BC066826.1 | - | 5 | 482 | |||
CN703950.1 | - | 5 | 561 |