Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145606.3 | NP_663581.1 | 7 | 1954 | ||
GenBank mRNA | AK046416.1 | - | 7 | 1927 | ||
AK080145.1 | - | 1 | 1052 | |||
AK080434.1 | - | 1 | 634 | |||
AK159830.1 | - | 7 | 1927 | |||
AK181175.1 | - | 1 | 137 | |||
AK196071.1 | - | 1 | 138 | |||
AK205507.1 | - | 1 | 137 | |||
AK216719.1 | - | 1 | 138 | |||
AK218446.1 | - | 1 | 138 | |||
BC010524.1 | - | 7 | 1905 | 3500937 | ||
BC023807.1 | - | 7 | 1917 | 5354266 | ||
BC036138.1 | - | 7 | 1906 | 4972627 | ||
BC036152.1 | - | 7 | 1908 | 4977509 | ||
CJ325786.1 | - | 1 | 243 |