Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145711.4 | NP_663757.3 | 7 | 1437 | ||
GenBank mRNA | AF472514.1 | - | 7 | 1233 | ||
AK029694.1 | - | 7 | 1234 | |||
AK038671.1 | - | 7 | 1440 | |||
AK133397.1 | - | 7 | 1234 | |||
AK161578.1 | - | 7 | 1223 | |||
BC038873.1 | - | 3 | 453 | |||
BC080732.1 | - | 7 | 1354 | 6518356 |