Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165954.1 | NP_001159426.1 | 13, 14 | 2631 | ||
NM_001165955.1 | NP_001159427.1 | 12, 13 | 2547 | |||
NM_001165956.1 | NP_001159428.1 | 14, 15 | 2505 | |||
NM_153421.2 | NP_700470.2 | 13, 14 | 2646 | |||
GenBank mRNA | AJ414610.1 | - | 13, 14 | 2632 | ||
AK037348.1 | - | 14, 15 | 2691 | |||
AK087339.1 | - | 12, 13 | 2541 | |||
AK166208.1 | - | 13, 14 | 2426 | |||
BC132533.1 | - | 13, 14 | 2615 | 40130810 | ||
BC145123.1 | - | 14, 15 | 2489 | 9053671 |