Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153422.2 | NP_700471.2 | 2 | 565 | ||
GenBank mRNA | AF541937.1 | - | 2 | 565 | ||
AK031275.1 | - | 2 | 791 | |||
BC119137.1 | - | 2 | 528 | 8733886 | ||
BC120486.1 | - | 2 | 528 | 40129409 |