Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nphp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153424.2 | NP_700473.2 | 2, 3 | 484 | ||
GenBank mRNA | AF537131.1 | - | 2, 3 | 438 | ||
AK016223.1 | - | 2, 3 | 449 | |||
AK038879.1 | - | 2, 3 | 239 | |||
AK084539.1 | - | 2, 3 | 484 | |||
AY118229.1 | - | 1, 2 | 176 | |||
BC138370.1 | - | 2, 3 | 209 | 8861390 |