Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15300
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
7
2320
NM_001272020.1
NP_001258949.1
6
2854
NM_004796.5
NP_004787.2
17
3683
NM_138970.4
NP_620426.2
6
2239
NR_073546.1
8
3632
NR_073547.1
21
6666
XM_005268218.2
XP_005268275.1
23
6047
XM_006720322.2
XP_006720385.1
22
6044
XM_006720323.2
XP_006720386.1
21
5708
XM_011537363.1
XP_011535665.1
23
6056
XM_011537364.2
XP_011535666.1
24
6018
XM_011537365.2
XP_011535667.1
23
5844
XM_011537366.1
XP_011535668.1
22
6032
XM_011537367.1
XP_011535669.1
22
5966
XM_011537368.1
XP_011535670.1
23
5753
XM_011537369.1
XP_011535671.1
23
5744
XM_011537370.1
XP_011535672.1
22
5720
XM_011537371.1
XP_011535673.1
23
5432
XM_011537372.1
XP_011535674.1
23
5423
XM_011537373.1
XP_011535675.1
22
5333
XM_011537377.2
XP_011535679.1
19
4348
XM_017021790.1
XP_016877279.1
24
6156
XM_017021791.1
XP_016877280.1
24
5939
XM_017021792.1
XP_016877281.1
22
5839
XM_017021793.1
XP_016877282.1
22
6029
XM_017021794.1
XP_016877283.1
21
6020
XM_017021795.1
XP_016877284.1
21
5815
XM_017021796.1
XP_016877285.1
21
5996
XM_017021797.1
XP_016877286.1
20
5996
XM_017021798.1
XP_016877287.1
20
5993
XM_017021799.1
XP_016877288.1
22
5957
XM_017021800.1
XP_016877289.1
22
5654
XM_017021801.1
XP_016877290.1
21
5627
XM_017021802.1
XP_016877291.1
20
5648
XM_017021803.1
XP_016877292.1
22
5402
XM_017021804.1
XP_016877293.1
22
5396
XM_017021805.1
XP_016877294.1
21
5387
XM_017021806.1
XP_016877295.1
21
5381
XM_017021807.1
XP_016877296.1
22
5342
GenBank mRNA
AB018286.1
-
17
3664
AJ316284.2
-
21
6454
AJ493127.1
-
8
2708
AK056530.1
-
6
2854
AK096247.1
-
1
845
AL833739.1
-
6
1534
BC059368.1
-
6
2165
BC068469.1
-
6
2165
BC142649.1
-
7
1753
BC150194.1
-
7
1753
BC152457.1
-
17
3664
8862590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline