Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430548M08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163760.1 | NP_001157232.1 | 12 | 1434 | ||
NM_001163761.1 | NP_001157233.1 | 11 | 1425 | |||
NM_172286.4 | NP_758490.3 | 12 | 1455 | |||
GenBank mRNA | AK032447.1 | - | 12 | 1456 | ||
AK044754.1 | - | 11 | 1264 | |||
AK046401.1 | - | 12 | 1457 | |||
AK122299.1 | - | 12 | 1441 | |||
AK133157.1 | - | 11 | 1423 | |||
AK172108.1 | - | 11 | 1426 | |||
BC027126.1 | - | 12 | 1434 | 5359842 |