Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430548M08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163760.1 | NP_001157232.1 | 13 | 1691 | ||
NM_001163761.1 | NP_001157233.1 | 12 | 1682 | |||
NM_172286.4 | NP_758490.3 | 13 | 1712 | |||
GenBank mRNA | AK032447.1 | - | 13 | 1713 | ||
AK044754.1 | - | 12 | 1521 | |||
AK046401.1 | - | 13 | 1714 | |||
AK122299.1 | - | 13 | 1698 | |||
AK133157.1 | - | 12 | 1680 | |||
AK172108.1 | - | 12 | 1683 | |||
BC027126.1 | - | 13 | 1691 | 5359842 |