Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spire2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172287.2 | NP_758491.1 | 5 | 935 | ||
GenBank mRNA | BC026502.1 | - | 4 | 763 | 4506974 | |
BC049152.1 | - | 5 | 935 | 6311485 | ||
BC054072.1 | - | 5 | 935 |