Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sulf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198565.1 | NP_001185494.1 | 4 | 602 | ||
NM_001198566.1 | NP_001185495.1 | 3 | 507 | |||
NM_172294.1 | NP_758498.1 | 4 | 586 | |||
GenBank mRNA | AK028285.1 | - | 3 | 406 | ||
AK037389.1 | - | 4 | 628 | |||
AK045002.1 | - | 4 | 501 | |||
AK078756.1 | - | 4 | 591 | |||
AK081929.1 | - | 3 | 409 | |||
AK129278.1 | - | 3 | 408 | |||
AY101178.1 | - | 4 | 611 | |||
BC034547.1 | - | 4 | 586 | 4500954 | ||
BC049276.1 | - | 4 | 586 | 4500954 | ||
CD348948.1 | - | 4 | 602 |