Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172300.3 | NP_758504.3 | 1, 2 | 419 | ||
GenBank mRNA | AK077793.1 | - | 1, 2 | 417 | ||
AK149770.1 | - | 1, 2 | 419 | |||
AK152529.1 | - | 1, 2 | 133 | |||
AK153328.1 | - | 1, 2 | 150 | |||
AK168942.1 | - | 1, 2 | 112 | |||
BC027266.1 | - | 1, 2 | 44 | |||
BC054412.1 | - | 1, 2 | 122 | 2646768 | ||
CA317477.1 | - | 1, 2 | 443 |