Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK040271.1 | - | 8, 9 | 1174 | ||
AK048843.1 | - | 8, 9 | 1340 | |||
AK146866.1 | - | 1365 | ||||
AK160810.1 | - | 8, 9 | 1254 | |||
BC060678.1 | - | 7, 8 | 1095 | |||
BC084587.1 | - | 8, 9 | 1269 | 3602481 | ||
EU380771.1 | - | 1391 | ||||
EU380772.1 | - | 1449 |