Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172748.2 | NP_766336.2 | 11 | 2768 | ||
GenBank mRNA | AK028405.1 | - | 11 | 2535 | ||
AK031431.1 | - | 9 | 2161 | |||
AK161937.1 | - | 2 | 739 | |||
AK162161.1 | - | 5 | 1147 | |||
BC053109.1 | - | 7 | 1761 | |||
BC059812.1 | - | 11 | 2768 | 6401846 |