Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankle1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172756.2 | NP_766344.2 | 8 | 1346 | ||
GenBank mRNA | AK049395.1 | - | 6 | 1653 | ||
AK078830.1 | - | 7 | 1576 | |||
AK080768.1 | - | 6 | 1659 | |||
AK083552.1 | - | 6 | 1645 | |||
AK087444.1 | - | 6 | 1664 | |||
BC030436.1 | - | 8 | 1285 | |||
BC094316.1 | - | 8 | 1303 | |||
BC145659.1 | - | 4 | 569 | |||
EU184014.1 | - | 8 | 1322 |