Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172760.3 | NP_766348.3 | 14, 15 | 1557 | ||
GenBank mRNA | AK037067.1 | - | 14, 15 | 1550 | ||
AK157875.1 | - | 1495 | ||||
BC018516.1 | - | 12, 13 | 1446 | 3155913 | ||
BC026617.1 | - | 9, 10 | 1502 | |||
BC058752.1 | - | 14, 15 | 1540 | 6485393 | ||
BC138887.1 | - | 14, 15 | 1514 | 8861909 |