Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mthfsd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166482.1 | NP_001159954.1 | 7 | 672 | ||
NM_172761.3 | NP_766349.1 | 7 | 766 | |||
GenBank mRNA | AK083317.1 | - | 7 | 706 | ||
AK141278.1 | - | 7 | 601 | |||
AK188342.1 | - | 2 | 73 | |||
BC023782.1 | - | 7 | 743 | |||
BC026652.1 | - | 6 | 642 | |||
BC052066.1 | - | 6 | 2049 | 5708660 |