Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sc5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172769.2 | NP_766357.1 | 6 | 1939 | ||
GenBank mRNA | AK043825.1 | - | 6 | 1898 | ||
AK077670.1 | - | 6 | 1897 | |||
AK154309.1 | - | 6 | 1902 |