Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc9a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172780.3 | NP_766368.2 | 9 | 1229 | ||
GenBank mRNA | AK032326.1 | - | 9 | 1210 | ||
AK122232.1 | - | 9 | 1200 | |||
BC130221.1 | - | 9 | 1214 | 40141414 |