Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adamts4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172845.2 | NP_766433.1 | 4 | 1548 | ||
GenBank mRNA | AF449483.1 | - | 1 | 41 | ||
AK028364.1 | - | 1549 | ||||
AK076295.1 | - | 3 | 923 | |||
AK083534.1 | - | 4 | 1548 | |||
AK156429.1 | - | 2 | 131 | |||
AK157287.1 | - | 1549 | ||||
AK164900.1 | - | 4 | 1544 | |||
AK173001.1 | - | 1426 | ||||
BC027773.1 | - | 4 | 1182 | 5345809 |