Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004855.4 | NP_004846.4 | 5 | 958 | ||
XM_005254795.4 | XP_005254852.1 | 3 | 387 | |||
XM_011522235.2 | XP_011520537.1 | 5 | 958 | |||
XM_011522236.2 | XP_011520538.1 | 4 | 853 | |||
XM_011522237.2 | XP_011520539.1 | 4 | 462 | |||
XM_017022730.1 | XP_016878219.1 | 4 | 377 | |||
XM_017022731.1 | XP_016878220.1 | 5 | 506 | |||
XM_017022732.1 | XP_016878221.1 | 3 | 387 | |||
GenBank mRNA | AK223330.1 | - | 5 | 663 | ||
AK293666.1 | - | 2 | 574 | |||
AK314706.1 | - | 5 | 933 | |||
BC017711.1 | - | 5 | 895 | 4430974 | ||
D42138.1 | - | 5 | 686 |