Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpy19l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172920.4 | NP_766508.3 | 19 | 1762 | ||
GenBank mRNA | AK031291.1 | - | 1 | 842 | ||
AK143845.1 | - | 20 | 1767 | |||
BC031382.1 | - | 12 | 940 | |||
BC116782.1 | - | 1 | 330 | |||
BC116784.1 | - | 1 | 330 |