- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: BC030500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173411.2 | NP_775587.1 | 3 | 1138 | ||
GenBank mRNA | BC030500.1 | - | 3 | 1148 | 5401612 |