Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173445.4 | NP_775621.2 | 7 | 1747 | ||
GenBank mRNA | AK030872.1 | - | 7 | 1718 | ||
AK047578.1 | - | 892 | ||||
AK084219.1 | - | 5 | 1673 | |||
AK088482.1 | - | 5 | 2130 | |||
AK088662.1 | - | 7 | 1388 | |||
BC069844.1 | - | 7 | 1727 | 6487223 |