Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175031.3 | NP_778196.2 | 6 | 740 | ||
GenBank mRNA | AK173142.1 | - | 6 | 700 | ||
BC043103.1 | - | 6 | 692 | 6406485 | ||
BC058698.1 | - | 6 | 805 | 6831702 | ||
BC061470.1 | - | 5 | 549 |