Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290570.1 | NP_001277499.1 | 3 | 410 | ||
NM_001290572.1 | NP_001277501.1 | 3 | 410 | |||
NM_175211.5 | NP_780420.1 | 3 | 410 | |||
GenBank mRNA | AK017939.1 | - | 3 | 386 | ||
AK076767.1 | - | 3 | 395 | |||
AK129123.1 | - | 3 | 377 | |||
AK170248.1 | - | 3 | 356 | |||
BC072656.1 | - | 3 | 345 | 30536315 | ||
CJ112478.1 | - | 3 | 412 |