Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prtg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175485.4 | NP_780694.3 | 5 | 951 | ||
GenBank mRNA | AK083540.1 | - | 3 | 387 | ||
AY630257.1 | - | 5 | 1021 | |||
BC116336.1 | - | 3 | 384 | |||
BC116337.1 | - | 3 | 384 | |||
DQ360114.1 | - | 5 | 951 |