Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204077.1 | NP_001191006.1 | 20 | 4845 | ||
NM_004788.3 | NP_004779.2 | 20 | 4866 | |||
GenBank mRNA | BC111417.1 | - | 20 | 4818 | 40038776 | |
BC112367.1 | - | 20 | 4821 | 40038767 | ||
D50916.1 | - | 20 | 4802 |