Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168561.1 | NP_001162033.1 | 2 | 586 | ||
NM_001168562.1 | NP_001162034.1 | 2 | 586 | |||
NM_175558.4 | NP_780767.1 | 2 | 480 | |||
GenBank mRNA | AK041761.1 | - | 2 | 482 | ||
AK044666.1 | - | 1 | 1321 | |||
AK142766.1 | - | 1 | 975 | |||
BC036728.1 | - | 1 | 432 | |||
BC048909.1 | - | 2 | 422 | |||
BC117850.1 | - | 2 | 593 | |||
BC117851.1 | - | 2 | 588 | 40093910 |