Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004795.3 | NP_004786.2 | 5 | 2850 | ||
XM_006719895.2 | XP_006719958.1 | 5 | 2274 | |||
GenBank mRNA | AB005142.1 | - | 5 | 2850 |