Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15392
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
7
2679
NM_001272020.1
NP_001258949.1
6
3213
NM_004796.5
NP_004787.2
17
4042
NM_138970.4
NP_620426.2
6
2598
NR_073546.1
8
3991
NR_073547.1
21
7025
XM_005268218.2
XP_005268275.1
23
6406
XM_006720322.2
XP_006720385.1
22
6403
XM_006720323.2
XP_006720386.1
21
6067
XM_011537363.1
XP_011535665.1
23
6415
XM_011537364.2
XP_011535666.1
24
6377
XM_011537365.2
XP_011535667.1
23
6203
XM_011537366.1
XP_011535668.1
22
6391
XM_011537367.1
XP_011535669.1
22
6325
XM_011537368.1
XP_011535670.1
23
6112
XM_011537369.1
XP_011535671.1
23
6103
XM_011537370.1
XP_011535672.1
22
6079
XM_011537371.1
XP_011535673.1
23
5791
XM_011537372.1
XP_011535674.1
23
5782
XM_011537373.1
XP_011535675.1
22
5692
XM_011537377.2
XP_011535679.1
19
4707
XM_017021790.1
XP_016877279.1
24
6515
XM_017021791.1
XP_016877280.1
24
6298
XM_017021792.1
XP_016877281.1
22
6198
XM_017021793.1
XP_016877282.1
22
6388
XM_017021794.1
XP_016877283.1
21
6379
XM_017021795.1
XP_016877284.1
21
6174
XM_017021796.1
XP_016877285.1
21
6355
XM_017021797.1
XP_016877286.1
20
6355
XM_017021798.1
XP_016877287.1
20
6352
XM_017021799.1
XP_016877288.1
22
6316
XM_017021800.1
XP_016877289.1
22
6013
XM_017021801.1
XP_016877290.1
21
5986
XM_017021802.1
XP_016877291.1
20
6007
XM_017021803.1
XP_016877292.1
22
5761
XM_017021804.1
XP_016877293.1
22
5755
XM_017021805.1
XP_016877294.1
21
5746
XM_017021806.1
XP_016877295.1
21
5740
XM_017021807.1
XP_016877296.1
22
5701
GenBank mRNA
AB018286.1
-
17
4023
AJ493127.1
-
8
3067
AK056530.1
-
6
3213
AK096247.1
-
1
1204
AL833739.1
-
6
1893
BC059368.1
-
6
2524
BC068469.1
-
6
2524
BC142649.1
-
7
2112
BC150194.1
-
7
2112
BC152457.1
-
17
4023
8862590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0