Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN1 SIRNA Assays ›
Gene Symbol:
NRXN1
Gene Name:
neurexin 1
Gene Aliases:
Hs.22998, PTHSL2, SCZD17
Chromosome Location:
Chr.2: 49918503 - 51032536 on Build GRCh38
UniGene ID:
Hs.637685
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15397
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001135659.1
NP_001129131.1
24
6251
NM_001320156.1
NP_001307085.1
3
762
NM_001320157.1
NP_001307086.1
3
753
NM_004801.4
NP_004792.1
22
6041
NM_138735.2
NP_620072.1
6
2266
XM_005264642.3
XP_005264699.1
23
6143
XM_005264643.3
XP_005264700.1
22
6098
XM_006712137.3
XP_006712200.1
22
6053
XM_011533167.2
XP_011531469.1
23
6152
XM_011533171.2
XP_011531473.1
23
6131
XM_011533172.2
XP_011531474.1
22
6125
XM_011533174.2
XP_011531476.1
22
6107
XM_011533175.2
XP_011531477.1
21
6095
XM_011533177.2
XP_011531479.1
22
6062
XM_011533178.2
XP_011531480.1
20
6062
XM_011533183.1
XP_011531485.1
17
3834
XM_017005303.1
XP_016860792.1
24
6179
XM_017005304.1
XP_016860793.1
24
6176
XM_017005305.1
XP_016860794.1
24
6170
XM_017005306.1
XP_016860795.1
23
6167
XM_017005307.1
XP_016860796.1
23
6161
XM_017005308.1
XP_016860797.1
24
6158
XM_017005309.1
XP_016860798.1
23
6152
XM_017005310.1
XP_016860799.1
22
6149
XM_017005311.1
XP_016860800.1
23
6134
XM_017005312.1
XP_016860801.1
23
6128
XM_017005313.1
XP_016860802.1
23
6122
XM_017005314.1
XP_016860803.1
21
6119
XM_017005315.1
XP_016860804.1
22
6116
XM_017005316.1
XP_016860805.1
22
6116
XM_017005317.1
XP_016860806.1
22
6110
XM_017005318.1
XP_016860807.1
22
6107
XM_017005319.1
XP_016860808.1
22
6104
XM_017005320.1
XP_016860809.1
22
6095
XM_017005321.1
XP_016860810.1
23
6089
XM_017005322.1
XP_016860811.1
23
6080
XM_017005323.1
XP_016860812.1
19
6065
XM_017005324.1
XP_016860813.1
21
6035
XM_017005325.1
XP_016860814.1
21
6026
XM_017005326.1
XP_016860815.1
21
6023
XM_017005327.1
XP_016860816.1
21
6008
XM_017005328.1
XP_016860817.1
20
5990
XM_017005330.1
XP_016860819.1
18
5966
XM_017005331.1
XP_016860820.1
18
5951
XM_017005332.1
XP_016860821.1
17
5930
XM_017005333.1
XP_016860822.1
17
5921
XM_017005334.1
XP_016860823.1
20
4000
XM_017005335.1
XP_016860824.1
7
7346
XM_017005336.1
XP_016860825.1
7
7338
XM_017005337.1
XP_016860826.1
6
7247
GenBank mRNA
AB011150.2
-
21
4804
AB035356.1
-
21
4756
AF064842.1
-
6
2265
AF087975.1
-
1
30
AK295773.1
-
3
656
BC125179.1
-
7
1322
BC125180.1
-
7
1685
BC150247.1
-
21
4825
9007472
BX647616.1
-
3
716
EF539882.1
-
24
6114
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
chemical synaptic transmission
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
vocal learning
neuromuscular process controlling balance
positive regulation of synapse assembly
positive regulation of synaptic transmission, glutamatergic
prepulse inhibition
regulation of insulin secretion involved in cellular response to glucose stimulus
vocalization behavior
positive regulation of synapse maturation
postsynaptic membrane assembly
gephyrin clustering involved in postsynaptic density assembly
neuroligin clustering involved in postsynaptic membrane assembly
postsynaptic density protein 95 clustering
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
receptor activity
calcium channel regulator activity
calcium ion binding
protein binding
acetylcholine receptor binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-8gqds:80/10.239.164.183:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0