Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301204.1 | NP_001288133.1 | 11 | 1595 | ||
NM_001301205.1 | NP_001288134.1 | 10 | 1280 | |||
NM_176913.4 | NP_795887.2 | 12 | 1752 | |||
GenBank mRNA | AF488552.1 | - | 12 | 1752 | ||
AK089244.1 | - | 11 | 1581 | |||
AK155417.1 | - | 11 | 1550 | |||
AK170970.1 | - | 11 | 1547 | |||
AK171044.1 | - | 11 | 1547 | |||
AK171122.1 | - | 11 | 1547 | |||
AK171155.1 | - | 10 | 1260 | |||
AK171268.1 | - | 11 | 1547 | |||
AK171503.1 | - | 11 | 1547 |