Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9330132A10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033982.1 | - | 3245 | |||
AK083197.1 | - | 3019 | ||||
BC098197.1 | - | 2972 |