Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THEMIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039477.2 | NP_001034566.1 | 3, 4 | 418 | ||
NM_001105556.2 | NP_001099026.1 | 5, 6 | 1902 | |||
NM_001286113.1 | NP_001273042.1 | 6, 7 | 1515 | |||
NM_001286115.1 | NP_001273044.1 | 5, 6 | 1314 | |||
NM_004848.3 | NP_004839.2 | 4, 5 | 829 | |||
XM_005246041.3 | XP_005246098.1 | 4, 5 | 1503 | |||
XM_006711050.1 | XP_006711113.1 | 6, 7 | 1461 | |||
XM_011542445.2 | XP_011540747.1 | 4, 5 | 1773 | |||
GenBank mRNA | AB035482.1 | - | 2, 3 | 374 | ||
AB050854.1 | - | 4, 5 | 1119 | |||
AF044896.1 | - | 3, 4 | 1556 | |||
AK094833.1 | - | 1, 2 | 2051 | |||
AK303090.1 | - | 6, 7 | 1477 | |||
AK303141.1 | - | 5, 6 | 1310 | |||
BC031655.1 | - | 4, 5 | 797 | |||
BC132692.1 | - | 4, 5 | 828 | |||
BC133049.1 | - | 4, 5 | 828 | |||
BG716895.1 | - | 3, 4 | 411 | |||
BI908174.1 | - | 2, 3 | 424 | |||
CR749321.1 | - | 3, 4 | 419 |