Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177059.3 | NP_796033.2 | 13, 14 | 1791 | ||
GenBank mRNA | AF374459.1 | - | 13, 14 | 1791 | ||
AK046350.1 | - | 13, 14 | 1842 | |||
AK081114.1 | - | 13, 14 | 1837 | |||
AK220367.1 | - | 9, 10 | 1171 | |||
BC132353.1 | - | 13, 14 | 1768 | 40130630 | ||
BC144824.1 | - | 13, 14 | 1761 |