Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ptpn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177081.3 | NP_796055.1 | 10 | 2449 | ||
GenBank mRNA | AK049175.1 | - | 9 | 2300 | ||
AK083305.1 | - | 10 | 2450 | |||
BC098475.1 | - | 10 | 2504 | 30044369 |