Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177353.3 | NP_796327.1 | 7 | 1013 | ||
GenBank mRNA | AK041169.1 | - | 7 | 1013 | ||
AK078577.1 | - | 7 | 1058 | |||
BC058750.1 | - | 7 | 1019 | 6390297 |