- ์ฃผ๋ฌธ ํํฉ
- ๋น ๋ฅธ์ฃผ๋ฌธ
-
ํ์์ด ์๋์ ๊ฐ์? ๊ณ์ ์์ฑํ๊ธฐ
Gene Symbol: Slc9a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177353.3 | NP_796327.1 | 9, 10 | 1321 | ||
GenBank mRNA | AK041169.1 | - | 9, 10 | 1321 | ||
BC058750.1 | - | 9, 10 | 1327 | 6390297 |