Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177364.3 | NP_796338.2 | 13 | 1293 | ||
GenBank mRNA | AK089330.1 | - | 13 | 1265 | ||
AK154898.1 | - | 13 | 1251 | |||
BC141306.1 | - | 13 | 1265 | 9055918 |