Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177715.4 | NP_808383.3 | 1 | 2050 | ||
GenBank mRNA | AK035535.1 | - | 1 | 2052 | ||
AK049420.1 | - | 1 | 175 | |||
AK172581.1 | - | 1 | 2051 | |||
AK173260.1 | - | 1 | 1555 | |||
BC116647.1 | - | 2 | 1437 | |||
BC116648.1 | - | 2 | 1437 |