Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178730.3 | NP_848845.1 | 3 | 260 | ||
GenBank mRNA | AK028447.1 | - | 3 | 262 | ||
AK028587.1 | - | 3 | 255 | |||
AK029032.1 | - | 3 | 222 | |||
AK037173.1 | - | 2 | 104 | |||
BC132629.1 | - | 2 | 110 | 40130906 |