Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egflam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178748.6 | NP_848863.1 | 17, 18 | 2778 | ||
GenBank mRNA | AK031136.1 | - | 17, 18 | 2687 | ||
AK033332.1 | - | 17, 18 | 2695 | |||
AK037223.1 | - | 17, 18 | 2680 | |||
AK041546.1 | - | 17, 18 | 2659 | |||
BC051455.1 | - | 3, 4 | 331 | |||
BC150710.1 | - | 17, 18 | 2498 | 9087621 |