Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178869.4 | NP_849200.2 | 11 | 1412 | ||
GenBank mRNA | AK031917.1 | - | 11 | 1383 | ||
AK047046.1 | - | 11 | 1397 | |||
AK140472.1 | - | 11 | 1380 | |||
AK168786.1 | - | 11 | 1383 | |||
AK169150.1 | - | 11 | 1399 | |||
BC010510.1 | - | 11 | 1361 | 4191464 |