Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bahcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198423.3 | NP_940815.3 | 16 | 5235 | ||
GenBank mRNA | AB257855.1 | - | 15 | 5237 | ||
AK147250.1 | - | 16 | 5235 | |||
AK147400.1 | - | 8 | 2143 | |||
BC072602.1 | - | 5 | 673 | 6830770 |