Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164491.1 | NP_001157963.1 | 5 | 611 | ||
NM_198649.3 | NP_941051.2 | 4 | 930 | |||
GenBank mRNA | AK086538.1 | - | 4 | 710 | ||
AK173040.1 | - | 4 | 488 | |||
BC060275.1 | - | 5 | 582 | 6822626 | ||
BC094229.1 | - | 4 | 904 | 30534236 |